AC5424074307 Accela ChemBio Inc.

3-Amino-3-(1-Boc-3-pyrrolidyl)propanoic Acid

  • MDL number MFCD06410589
  • Cas Number : 889949-27-3
  • Molecular Weight : 258.31408

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015841&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015841.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart