AC5424074308 Accela ChemBio Inc.

6-(3-Fluorophenyl)-2-pyridinecarboxylic Acid

  • MDL number MFCD06410053
  • Cas Number : 887982-40-3
  • Molecular Weight : 217.1958232

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015844&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY015844.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart