AC5424074325 Accela ChemBio Inc.

6-[2-(Trifluoromethoxy)phenyl]-3-pyridinecarbaldehyde

  • MDL number MFCD04115434
  • Cas Number : 898405-31-7
  • Molecular Weight : 267.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015945&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY015945.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart