AC5424074333 Accela ChemBio Inc.

[6-(3,5-Dichlorophenyl)-3-pyridyl]methanol

  • MDL number MFCD06410460
  • Cas Number : 887974-84-7
  • Molecular Weight : 254.11196

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015963&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY015963.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart