AC5424074344 Accela ChemBio Inc.

2-Mercapto-6,7-dihydro-3H-cyclopentapyrimidin-4(5H)-one

  • MDL number MFCD02355518
  • Cas Number : 35563-27-0
  • Molecular Weight : 168.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016087&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY016087.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart