AC5424074367 Accela ChemBio Inc.

(4-Amino-4-tetrahydrothiopyranyl)methanol

  • MDL number MFCD04115579
  • Cas Number : 898796-23-1
  • Molecular Weight : 147.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016240&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016240.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart