AC5424074371 Accela ChemBio Inc.

6-[4-(Trifluoromethoxy)phenyl]-2-pyridinecarbaldehyde

  • MDL number MFCD06410031
  • Cas Number : 887979-25-1
  • Molecular Weight : 267.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016249&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY016249.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart