AC5424074392 Accela ChemBio Inc.

2-(4-Iodophenyl)-3-(2,4-dinitrophenyl)-5-(2,4-disulfophenyl)-2H-tetrazolium Sodium Salt

  • MDL number MFCD01075080
  • Cas Number : 515111-36-1
  • Molecular Weight : 696.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016316&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY016316.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart