AC5424074402 Accela ChemBio Inc.

1-(Boc-amino)-1,2,3,4-tetrahydronaphthalene-1-carboxylic Acid

  • MDL number MFCD04115523
  • Cas Number : 898404-93-8
  • Molecular Weight : 291.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016368&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY016368.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart