AC5424074406 Accela ChemBio Inc.

5-[3,5-Bis(trifluoromethyl)phenyl]-3-pyridinecarbaldehyde

  • MDL number MFCD06410252
  • Cas Number : 887973-75-3
  • Molecular Weight : 319.2018992

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016378&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY016378.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart