AC5424074492 Accela ChemBio Inc.

3-Oxo-2,3-dihydropyridazine-4-carboxamide

  • MDL number MFCD19208481
  • Cas Number : 2125-92-0
  • Molecular Weight : 139.1121

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017202&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY017202.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart