AC5424074493 Accela ChemBio Inc.

4-(4-Nitrophenyl)-2-piperazinone

  • MDL number MFCD18916775
  • Cas Number : 223785-99-7
  • Molecular Weight : 221.21264

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017203&attname=GoodsCode

Safety Information

hcodes No GHS SYMBOl
MSDS http://pdf.accelachem.com/cn/msds/SY017203.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart