AC5424074494 Accela ChemBio Inc.

Methyl cis-1-Boc-4-aminopiperidine-3-carboxylate

  • MDL number MFCD21332897
  • Cas Number : 1620128-54-2
  • Molecular Weight : 258.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017204&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY017204.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart