AC5424074495 Accela ChemBio Inc.

cis-1-Boc-4-amino-3-piperidinemethanol

  • MDL number MFCD21332898
  • Cas Number : 1402249-02-8
  • Molecular Weight : 230.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017205&attname=GoodsCode

Safety Information

hcodes No GHS SYMBOl
MSDS http://pdf.accelachem.com/cn/msds/SY017205.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart