AC5424074518 Accela ChemBio Inc.

Ethyl 2-Oxocycloheptanecarboxylate

  • MDL number MFCD00604086
  • Cas Number : 774-05-0
  • Molecular Weight : 184.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017443&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY017443.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart