AC5424074614 Accela ChemBio Inc.

6-Methoxyquinazoline-2,4(1H,3H)-dione

  • MDL number MFCD09954838
  • Cas Number : 32618-84-1
  • Molecular Weight : 192.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methoxy-2,4-quinazolinediol
Inchi Code 1S/C9H8N2O3/c1-14-5-2-3-7-6(4-5)8(12)11-9(13)10-7/h2-4H,1H3,(H2,10,11,12,13)
Inchi Key KAYIYDVQBAPJIU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018575&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY018575.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart