AC5424074625 Accela ChemBio Inc.

2-[(Ethylamino)methyl]phenol

  • MDL number MFCD09929100
  • Cas Number : 108206-05-9
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018670&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY018670.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart