AC5424074629 Accela ChemBio Inc.

Ethyl 1-Boc-3-allylpiperidine-3-carboxylate

  • MDL number MFCD11977500
  • Cas Number : 374795-32-1
  • Molecular Weight : 297.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018696&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY018696.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart