AC5424074635 Accela ChemBio Inc.

(3-Chloropropyl)sulfamoyl Chloride

  • MDL number MFCD14529128
  • Cas Number : 42065-72-5
  • Molecular Weight : 192.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018716&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY018716.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart