AC5424074651 Accela ChemBio Inc.

2-Ethyloxazolo[5,4-b]pyridine

  • MDL number MFCD18384507
  • Cas Number : 856990-30-2
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018741&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY018741.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart