AC5424074668 Accela ChemBio Inc.

4-Hydroxy-2-(trifluoromethyl)benzoic Acid

  • MDL number MFCD01091007
  • Cas Number : 320-32-1
  • Molecular Weight : 206.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018811&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501A
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY018811.pdf
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart