AC5424074670 Accela ChemBio Inc.

Benzyl 2-Methoxy-2-methylpropionate

  • MDL number MFCD21641338
  • Cas Number : 1363165-96-1
  • Molecular Weight : 208.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018847&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018847.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart