AC5424074687 Accela ChemBio Inc.

Methyl 5-(Chlorosulfonyl)pentanoate

  • MDL number MFCD22574974
  • Cas Number : 1408058-14-9
  • Molecular Weight : 214.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019028&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY019028.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart