AC5424074699 Accela ChemBio Inc.

Methyl 2-Isopropylbenzoate

  • MDL number MFCD20484275
  • Cas Number : 6623-98-9
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019141&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY019141.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart