AC5424074729 Accela ChemBio Inc.

4-(Chloromethyl)phenylacetic Acid

  • MDL number MFCD04038831
  • Cas Number : 56066-91-2
  • Molecular Weight : 184.61956

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019601&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY019601.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart