AC5424074731 Accela ChemBio Inc.

6-Hydroxybenzofuran-2-carboxylic Acid

  • MDL number MFCD11101030
  • Cas Number : 334022-87-6
  • Molecular Weight : 178.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019648&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY019648.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart