AC5424074768 Accela ChemBio Inc.

Methyl 4-Morpholino-2-nitrobenzoate

  • MDL number MFCD22205882
  • Cas Number : 404010-97-5
  • Molecular Weight : 266.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019798&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY019798.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart