AC5424074780 Accela ChemBio Inc.

4-[(Diethylamino)methyl]phenylboronic Acid

  • MDL number MFCD04039019
  • Cas Number : 220999-48-4
  • Molecular Weight : 207.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019883&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019883.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart