AC5424074866 Accela ChemBio Inc.

1-Boc-2-piperidinamide

  • MDL number MFCD02183582
  • Cas Number : 388077-74-5
  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY020462&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY020462.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart