AC5424074932 Accela ChemBio Inc.

Diethyl 2-Fluorobenzylphosphonate

  • MDL number MFCD22574982
  • Cas Number : 63909-54-6
  • Molecular Weight : 246.2151042

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021333&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY021333.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart