AC5424074934 Accela ChemBio Inc.

Diethyl 3-Nitrobenzylphosphonate

  • MDL number MFCD22574984
  • Cas Number : 104097-04-3
  • Molecular Weight : 273.222201

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021339&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021339.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart