AC5424074940 Accela ChemBio Inc.

2-Bromo-5-fluorobenzyl Chloride

  • MDL number MFCD09263441
  • Cas Number : 857276-61-0
  • Molecular Weight : 223.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021377&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY021377.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart