AC5424074999 Accela ChemBio Inc.

3-(2-Pyridyl)-2-propyn-1-ol

  • MDL number MFCD00168870
  • Cas Number : 29768-03-4
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022166&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY022166.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart