AC5424075000 Accela ChemBio Inc.

3-(4-Pyridyl)-2-propen-1-ol

  • MDL number MFCD18828038
  • Cas Number : 193002-34-5
  • Molecular Weight : 135.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022167&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY022167.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart