AC5424075005 Accela ChemBio Inc.

(2-Fluorophenyl)trimethylsilane

  • MDL number MFCD00092633
  • Cas Number : 1842-26-8
  • Molecular Weight : 168.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022241&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY022241.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart