AC5424075024 Accela ChemBio Inc.

4-(2-Aminoethyl)-4H-1,2,4-triazole

  • MDL number MFCD03093052
  • Cas Number : 1008526-84-8
  • Molecular Weight : 112.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022380&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022380.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart