AC5424075025 Accela ChemBio Inc.

Methyl 5-Bromo-1H-1,2,3-triazole-4-carboxylate

  • MDL number MFCD23381099
  • Cas Number : 1427475-25-9
  • Molecular Weight : 206

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022386&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY022386.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart