AC5424075033 Accela ChemBio Inc.

3-Benzyl-3-azabicyclo[3.2.1]octan-8-one

  • MDL number MFCD08275096
  • Cas Number : 83507-33-9
  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022397&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY022397.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart