AC5424075047 Accela ChemBio Inc.

3-(3-Pyridyl)propanamide

  • MDL number MFCD00956246
  • Cas Number : 84199-99-5
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022419&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY022419.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart