AC5424075058 Accela ChemBio Inc.

Ethyl 3-(2-Pyridyl)propanoate

  • MDL number MFCD04004149
  • Cas Number : 2739-74-4
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022461&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022461.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart