AC5424075063 Accela ChemBio Inc.

Methyl 8-Isoquinolinecarboxylate

  • MDL number MFCD11506141
  • Cas Number : 850858-56-9
  • Molecular Weight : 187.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022478&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY022478.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart