AC5424075067 Accela ChemBio Inc.

1-Methyl-4-benzimidazolecarboxylic Acid

  • MDL number MFCD09751736
  • Cas Number : 672957-92-5
  • Molecular Weight : 176.17202

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022483&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY022483.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart