AC5424075101 Accela ChemBio Inc.

N-(3-Indolylmethyl)benzylamine

  • MDL number MFCD17430949
  • Cas Number : 57506-64-6
  • Molecular Weight : 236.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022651&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY022651.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart