AC5424075127 Accela ChemBio Inc.

4,5-Dimethylthiazole-2-carboxylic Acid

  • MDL number MFCD11934399
  • Cas Number : 79312-41-7
  • Molecular Weight : 157.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY022801&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY022801.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart