AC5424075151 Accela ChemBio Inc.

4-Ethoxyphenylacetic Acid

  • MDL number MFCD00004346
  • Cas Number : 4919-33-9
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023161&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501A
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY023161.pdf
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart