AC5424075155 Accela ChemBio Inc.

Ethyl 4-Chlorobenzoate

  • MDL number MFCD00013645
  • Cas Number : 7335-27-5
  • Molecular Weight : 184.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023174&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P362-P403+P233-P501c
hcodes GHS07,
MSDS http://pdf.accelachem.com/cn/msds/SY023174.pdf
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart