AC5424075168 Accela ChemBio Inc.

4-Nitrophthalide

  • MDL number MFCD03069001
  • Cas Number : 65399-18-0
  • Molecular Weight : 179.1296

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023223&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY023223.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart