AC5424075197 Accela ChemBio Inc.

7-Bromo-6-fluoroisoquinoline

  • MDL number MFCD11846300
  • Cas Number : 923022-40-6
  • Molecular Weight : 226.0451032

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023347&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY023347.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart