AC5424075201 Accela ChemBio Inc.

3,3-Pentamethylene Glutarimide

  • MDL number MFCD00023872
  • Cas Number : 1130-32-1
  • Molecular Weight : 181.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023364&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY023364.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart