AC5424075226 Accela ChemBio Inc.

2-(Boc-amino)phenylboronic Acid Pinacol Ester

  • MDL number MFCD03411943
  • Cas Number : 159624-15-4
  • Molecular Weight : 319.20364

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023463&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY023463.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart